Imidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Boiling Pt:235 °C
Melting Pt:85…91 °C
Flash Pt:145 °C
Storage Temperature:Ambient
EINECS:206-019-2
UN:3263
ADR:8,II
:68.08 g/mol
:GHS05
:GHS07
:GHS08
Frequently Bought Together