
2-Bromobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:88-65-3
:
Formula:C₇H₅BrO₂
MDL Number:MFCD00002402
:201.02 g/mol
Melting Pt:146…150 °C
Density:1,93 g/cm³ (25 °C)
EINECS:201-848-6
:GHS07
Storage Temperature:Ambiente
Frequently Bought Together