Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
 

1,1-Dimethylurea

Irritant
Supplier:   Apollo Scientific
1,1-Dimethylurea
Supplier:  Apollo Scientific
Certificates
1,1-Dimethylurea

2 Options Available Below

Product Details & Documents
Synonyms:  N,N-Dimethylurea
  • CAS Number:598-94-7
  • Formula:C₃H₈N₂O
  • MDL Number:MFCD00007959
  • Molecular Weight:88.11 g/mol
  • Melting Pt:178…183 °C
  • Density:1,255 g/cm³ (25 °C)
  • Storage Temperature:Ambiente
Product Family Options

Product Information

  • Tipo di conf.
    Conf
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,20 €
    36,20 €
    1 * 25 g
       36,20 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 138,00 €
    138,00 €
    1 * 100 g
       138,00 €