Order Entry
Italy
ContactUsLinkComponent
(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine 95%
(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine 95%
Catalog # APOSOR480852-1G
Supplier:  Apollo Scientific
CAS Number:  
(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine 95%
Catalog # APOSOR480852-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :139.2 g/mol
Storage Temperature:Ambient
CAS Number:848436-19-1
Formula:C₇H₁₃N₃
MDL Number:MFCD02090911