Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-1H-pyrrolo[3,2-b]pyridine
5-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
5-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
APOSOR480796-5G
:
Apollo Scientific
:
5-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
APOSOR480796-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
152.5800 g/mol
:
Ambient
:
65156-94-7
:
C₇H₅ClN₂
:
MFCD08062553