2,3-Dichlorobenzenesulphonamide 97%
Catalog # APOSOR480709-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:226.08 g/mol
Storage Temperature:Ambient
CAS Number:82967-94-0
Formula:C₆H₅Cl₂NO₂S
MDL Number:MFCD00464303