Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Methoxy-1-naphthaldehyde 95%

4-Methoxy-1-naphthaldehyde 95%

Supplier:   Apollo Scientific
4-Methoxy-1-naphthaldehyde 95%
Supplier:  Apollo Scientific
Certificates
4-Methoxy-1-naphthaldehyde 95%

3 Options Available Below

Product Details & Documents
Synonyms:  4-Methoxynaphthalene-1-carbaldehyde
  • CAS Number:15971-29-6
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00004006
  • Molecular Weight:186.21 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 57,90 €
    57,90 €
    1 * 25 g
       57,90 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 222,00 €
    222,00 €
    1 * 100 g
       222,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.