



Ethoxyacetylene 50% in hexane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:927-80-0
:
Formula:C₄H₆O
MDL Number:MFCD00009247
:70.091 g/mol
Boiling Pt:55…56 °C
Density:0.718 g/cm³ (25 °C)
Flash Pt:–28 °C
EINECS:213-164-5
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Frequently Bought Together