Order Entry
Italy
ContactUsLinkComponent
Nortropinone hydrochloride 97%
Irritant
Nortropinone hydrochloride 97%
Catalog # APOSOR4717-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Nortropinone hydrochloride 97%
Catalog # APOSOR4717-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :161.63 g/mol
Melting Pt:204 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:25602-68-0
Formula:C₇H₁₂ClNO
MDL Number:MFCD03613582