Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Chlorothioxanthone 98%

Supplier:   Apollo Scientific
2-Chlorothioxanthone 98%
Supplier:  Apollo Scientific
Certificates
2-Chlorothioxanthone 98%

4 Options Available Below

Product Details & Documents
Synonyms:  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  • CAS Number:86-39-5
  • Formula:C₁₃H₇ClOS
  • MDL Number:MFCD00005067
  • Molecular Weight:246.72 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 50,70 €
    50,70 €
    1 * 100 g
       50,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 150,00 €
    1 150,00 €
    1 * 2,5 kg
       1 150,00 €

    Specifications:
    New Product
    New Product