(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:209482-27-9
:
Formula:C₃₆H₃₀NO₂P
MDL Number:MFCD04117688
:539.61 g/mol
Storage Temperature:Ambient
Frequently Bought Together