Order Entry
Italy
Orders LinkContactUsLinkComponent
2,5-Dimethyl-p-benzoquinone

2,5-Dimethyl-p-benzoquinone

Supplier:   Apollo Scientific
2,5-Dimethyl-p-benzoquinone
Supplier:  Apollo Scientific
Certificates
2,5-Dimethyl-p-benzoquinone

4 Options Available Below

Product Details & Documents
Synonyms:  2,5-Dimethylcyclohexa-2,5-diene-1,4-dione
  • CAS Number:137-18-8
  • Formula:C₈H₈O₂
  • MDL Number:MFCD00041737
  • Molecular Weight:136.15 g/mol
  • Melting Pt:123…125 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 123,00 €
    123,00 €
    1 * 5 g
       123,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 497,00 €
    497,00 €
    1 * 25 g
       497,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 310,00 €
    1 310,00 €
    1 * 100 g
       1 310,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.