
2-Formyl-4-methoxypyridine
Catalog # APOSOR46108-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.1400 g/mol
Boiling Pt:125…130 °C/25mmHg
Melting Pt:28…30 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:16744-81-3
Formula:C₇H₇NO₂
MDL Number:MFCD08234865