

2-Methyl-1,3-dioxolane
:497-26-7
:
:C₄H₈O₂
:MFCD00003210
:88.11 g/mol
:83 °C/760mmHg
:0.982 g/cm³ (25 °C)
:–2 °C
:Ambient
:1993
:3,II
:GHS02
:GHS07
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Single-use syringes, 2-piece, HENKE-JECT®
: HENKE SASS WOLF