Order Entry
Italy
ContactUsLinkComponent
(R)-(-)-3-Amino-1-boc-piperidine
Irritant
(R)-(-)-3-Amino-1-boc-piperidine
Catalog # APOSOR4589-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(R)-(-)-3-Amino-1-boc-piperidine
Catalog # APOSOR4589-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :200.28 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:188111-79-7
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD03094717