1-(Tetrahydro-2H-pyran-4-yl)piperidin-4-amine dihydrochloride 97%
Catalog # APOSOR452210-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Amino-1-(oxan-4-yl)piperidine dihydrochloride
:
:257.21 g/mol
Storage Temperature:Ambient
CAS Number:794471-13-9
:755039-78-2
Formula:C₁₀H₂₂Cl₂N₂O
MDL Number:MFCD09759192