Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
1,6-Hexanedithiol 97+%

1,6-Hexanedithiol 97+%

Irritant
Supplier:   Apollo Scientific
1,6-Hexanedithiol 97+%
Supplier:  Apollo Scientific
Certificates
1,6-Hexanedithiol 97+%

3 Options Available Below

Product Details & Documents
Synonyms:  1,6-Dimercaptohexane
  • CAS Number:1191-43-1
  • Formula:C₆H₁₄S₂
  • MDL Number:MFCD00004910
  • Molecular Weight:150.31 g/mol
  • Boiling Pt:118…119 °C
  • Melting Pt:–21 °C
  • Density:0.995 g/cm³ (25 °C)
  • Flash Pt:91 °C
  • Storage Temperature:Ambient
  • UN:3334
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 29,00 €
    29,00 €
    1 * 25 g
       29,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 86,90 €
    86,90 €
    1 * 100 g
       86,90 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 304,00 €
    304,00 €
    1 * 500 g
       304,00 €