
(2-Mercaptoethyl)pyrazine 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35250-53-4
:
Formula:C₆H₈N₂S
MDL Number:MFCD00053160
:140.20 g/mol
Boiling Pt:105…110 °C
Density:1.142 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS06
Frequently Bought Together