Order Entry
Italy
ContactUsLinkComponent
7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine
7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine
Catalog # APOSOR451245-1G
Supplier:  Apollo Scientific
CAS Number:  
7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine
Catalog # APOSOR451245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.17 g/mol
Storage Temperature:Ambient
CAS Number:1211486-21-3
Formula:C₇H₉N₃O
MDL Number:MFCD15202267