Order Entry
Italy
ContactUsLinkComponent
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Corrosive
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Catalog # APOSOR43634-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Catalog # APOSOR43634-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :139.58 g/mol
Melting Pt:144…146 °C
 :GHS05
Storage Temperature:Ambient
CAS Number:623-33-6
Formula:C₄H₁₀ClNO₂
MDL Number:MFCD00012871