Order Entry
Italy
Orders LinkContactUsLinkComponent
1,6-Naphthyridine

1,6-Naphthyridine

Supplier:   Apollo Scientific
1,6-Naphthyridine
Supplier:  Apollo Scientific
Certificates
1,6-Naphthyridine

4 Options Available Below

Product Details & Documents
Synonyms:  Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene
  • CAS Number:253-72-5
  • Formula:C₈H₆N₂
  • MDL Number:MFCD00059750
  • Molecular Weight:130.15 g/mol
  • Melting Pt:35 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 33,30 €
    33,30 €
    1 * 100 mg
       33,30 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 261,00 €
    261,00 €
    1 * 1 g
       261,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 210,00 €
    1 210,00 €
    1 * 5 g
       1 210,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 5 780,00 €
    5 780,00 €
    1 * 25 g
       5 780,00 €

    Specifications:
    New Product
    New Product