Order Entry
Italy
Orders LinkContactUsLinkComponent
5-Bromo-3-nitro-2-picoline 97%

5-Bromo-3-nitro-2-picoline 97%

Supplier:   Apollo Scientific
5-Bromo-3-nitro-2-picoline 97%
Supplier:  Apollo Scientific
Certificates
5-Bromo-3-nitro-2-picoline 97%

4 Options Available Below

Product Details & Documents
Synonyms:  5-Bromo-2-methyl-3-nitropyridine
  • CAS Number:911434-05-4
  • Formula:C₆H₅BrN₂O₂
  • MDL Number:MFCD09031419
  • Molecular Weight:217.02 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 63,70 €
    63,70 €
    1 * 25 g
       63,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 170,00 €
    1 170,00 €
    1 * 500 g
       1 170,00 €

    Specifications:
    New Product
    New Product