
Succinic anhydride 97%
Catalog # APOSOR42120-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:100.07 g/mol
Boiling Pt:261 °C
Melting Pt:118…120 °C
Density:1.572 g/cm³ (25 °C)
Flash Pt:157 °C
Storage Temperature:Ambient
EINECS:203-570-0
:GHS07
CAS Number:108-30-5
Formula:C₄H₄O₃
MDL Number:MFCD00005525