Order Entry
Italy
ContactUsLinkComponent
1-(1-Naphthylmethyl)piperazine 97%
Irritant
Environment
1-(1-Naphthylmethyl)piperazine 97%
Catalog # APOSOR4166-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
1-(1-Naphthylmethyl)piperazine 97%
Catalog # APOSOR4166-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :226.323 g/mol
Melting Pt:63…67 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:40675-81-8
Formula:C₁₅H₁₈N₂
MDL Number:MFCD01314185