Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-morpholine-2-carboxylic acid
N-Boc-morpholine-2-carboxylic acid, Technical Grade
N-Boc-morpholine-2-carboxylic acid, Technical Grade
APOSOR41179-1G
:
Apollo Scientific
:
N-Boc-morpholine-2-carboxylic acid, Technical Grade
APOSOR41179-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
231.2500 g/mol
:
GHS07
:
Ambient
:
189321-66-2
:
C₁₀H₁₇NO₅
:
MFCD01321006