Order Entry
Italy
Orders LinkContactUsLinkComponent
1,1-Dimethoxyacetone

1,1-Dimethoxyacetone

Flammable
Supplier:   Apollo Scientific
1,1-Dimethoxyacetone
Supplier:  Apollo Scientific
Certificates
1,1-Dimethoxyacetone

3 Options Available Below

Product Details & Documents
Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:133…135 °C
  • Melting Pt:–57 °C
  • Density:1 g/cm³ (25 °C)
  • Flash Pt:37 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 98,40 €
    98,40 €
    1 * 500 g
       98,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 468,00 €
    468,00 €
    1 * 2,5 kg
       468,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 870,00 €
    1 870,00 €
    1 * 10 kg
       1 870,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.