
2-Chloro-p-cresol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6640-27-3
:
Formula:C₇H₇ClO
MDL Number:MFCD00070639
Boiling Pt:196 °C
Density:1.21 g/cm³ (25 °C)
Flash Pt:86 °C
Storage Temperature:Ambient
UN:2669
ADR:6.1,II
:142.58 g/mol
:GHS06
Frequently Bought Together