Order Entry
Italy
ContactUsLinkComponent
4-Bromo-2,5-xylidine 96%
Irritant
4-Bromo-2,5-xylidine 96%
Catalog # APOSOR3788-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-2,5-xylidine 96%
Catalog # APOSOR3788-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.0757 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30273-40-6
Formula:C₈H₁₀BrN
MDL Number:MFCD07780642