1,1-Dichloroethane 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Ethylidene dichloride
- CAS Number:75-34-3
- Formula:Cl₂HCCH₃
- MDL Number:MFCD00013673
- Molecular Weight:98.96 g/mol
- Boiling Pt:57 °C
- Melting Pt:–98 °C
- Density:1.18 g/cm³ (25 °C)
- Flash Pt:–10 °C
- Storage Temperature:Ambient
- EINECS:200-863-5
- UN:2362
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: Ethylidene dichloride
- CAS Number:75-34-3
- Formula:Cl₂HCCH₃
- MDL Number:MFCD00013673
- Molecular Weight:98.96 g/mol
- Boiling Pt:57 °C
- Melting Pt:–98 °C
- Density:1.18 g/cm³ (25 °C)
- Flash Pt:–10 °C
- Storage Temperature:Ambient
- EINECS:200-863-5
- UN:2362
- ADR:3,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



