Order Entry
Italy
ContactUsLinkComponent
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
Catalog # APOSOR361686-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
Catalog # APOSOR361686-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester
 :
 :288.96 g/mol
Storage Temperature:Ambient
CAS Number:1003298-87-0
Formula:C₁₂H₁₅BCl₂O₃
MDL Number:MFCD20526385