Order Entry
Italy
ContactUsLinkComponent
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Catalog # APOSOR361570-1G
Supplier:  Apollo Scientific
CAS Number:  
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Catalog # APOSOR361570-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :454.18 g/mol
Storage Temperature:Refrigerator
CAS Number:688756-58-3
Formula:C₂₈H₃₂B₂O₄
MDL Number:MFCD15143601