1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:70356-09-1
:
Formula:C₂₀H₂₂O₃
MDL Number:MFCD00210252
:310.39 g/mol
Storage Temperature:Ambient
Frequently Bought Together