
trans-4-(Aminomethyl)cyclohexanecarboxylic acid 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1197-18-8
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00001466
:157.21 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together