Sodium-6,7-dihydroxynaphthalene-2-sulphonate
Catalog # APOSOR350345-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:262.21 g/mol
Storage Temperature:Ambient
CAS Number:135-53-5
Formula:C₁₀H₈O₅S
MDL Number:MFCD00004074