Order Entry
Italy
ContactUsLinkComponent
1-Methylazepan-4-one hydrochloride
1-Methylazepan-4-one hydrochloride
Catalog # APOSOR350265-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methylazepan-4-one hydrochloride
Catalog # APOSOR350265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.6452 g/mol
Storage Temperature:Ambient
CAS Number:19869-42-2
Formula:C₇H₁₄ClNO
MDL Number:MFCD07779465