
3-Formyl-6-methoxypyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65873-72-5
:
Formula:C₇H₇NO₂
MDL Number:MFCD02683446
:137.14 g/mol
Melting Pt:49…54 °C
Flash Pt:107 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together