5-Bromo-2-hydroxy-3-nitro-4-picoline 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Bromo-2-hydroxy-4-methyl-3-nitropyridine, 5-bromo-2-hydroxy-3-nitro-4-picoline
CAS Number:228410-90-0
:
Formula:C₆H₅BrN₂O₃
MDL Number:MFCD06659520
:233.0200 g/mol
Melting Pt:240…243 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together