3-(6-Methylpyridin-2-yl)propan-1-ol
Catalog # APOSOR346269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.21 g/mol
Storage Temperature:Ambient
CAS Number:61744-43-2
Formula:C₉H₁₃NO
MDL Number:MFCD00012344