Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Aminomethyl)-2-bromopyridine
5-(Aminomethyl)-2-bromopyridine 98%
5-(Aminomethyl)-2-bromopyridine 98%
APOSOR345226-250MG
:
Apollo Scientific
:
5-(Aminomethyl)-2-bromopyridine 98%
APOSOR345226-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
187.04 g/mol
:
GHS07
:
Ambient
:
120740-10-5
:
C₆H₇BrN₂
:
MFCD06738744