Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide 95%
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide 95%
APOSOR33557-1G
:
Apollo Scientific
:
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide 95%
APOSOR33557-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
153.19 g/mol
:
Ambient
:
40639-97-2
:
C₇H₁₁N₃O
:
MFCD09909729