2-Chloro-5-(1H-pyrazol-1-ylmethyl)-1,3-thiazole 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:453557-78-3
:
Formula:C₇H₆ClN₃S
MDL Number:MFCD01871711
:199.66 g/mol
Storage Temperature:Ambient
Frequently Bought Together