4-(1,4-Dioxa-8-azaspiro[4.5]dec-8-yl)benzenecarbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:79421-40-2
:
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD06659581
:247.3 g/mol
Storage Temperature:Ambient
Frequently Bought Together