5-Methoxy-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26990-71-6
:
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD03791232
:154.17 g/mol
Storage Temperature:Ambient
Frequently Bought Together