Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77156-75-3
:
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD00173390
:231.2500 g/mol
Storage Temperature:Ambient
Frequently Bought Together