Order Entry
Italy
ContactUsLinkComponent
Morpholin-4-yl(pyridin-3-yl)acetonitrile 95%
Morpholin-4-yl(pyridin-3-yl)acetonitrile 95%
Catalog # APOSOR32849-1G
Supplier:  Apollo Scientific
CAS Number:  
Morpholin-4-yl(pyridin-3-yl)acetonitrile 95%
Catalog # APOSOR32849-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.25 g/mol
Storage Temperature:Ambient
CAS Number:36740-09-7
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD00215318