Order Entry
Italy
ContactUsLinkComponent
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate 95%
Catalog # APOSOR32821-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate 95%
Catalog # APOSOR32821-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.19 g/mol
Storage Temperature:Ambient
CAS Number:85795-19-3
Formula:C₉H₁₁NO₃
MDL Number:MFCD00141903