1-(3-Chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid
:339029-56-0
:
:C₁₁H₇ClN₂O₄
:MFCD00141204
:266.64 g/mol
:Ambient
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals