(R)-4-Hydroxypyrrolidin-2-one
Catalog # APOSOR322659-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:101.11 g/mol
Storage Temperature:Ambient
CAS Number:22677-21-0
Formula:C₄H₇NO₂
MDL Number:MFCD00273364