Order Entry
Italy
ContactUsLinkComponent
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
Catalog # APOSOR322627-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
Catalog # APOSOR322627-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.2 g/mol
Melting Pt:74…77 °C
Storage Temperature:Refrigerator
CAS Number:1005-28-3
Formula:C₆H₈N₂OS
MDL Number:MFCD03844445