Order Entry
Italy
ContactUsLinkComponent
 
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-amine 95%
Catalog # APOSOR32245-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-amine 95%
Catalog # APOSOR32245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.2 g/mol
Storage Temperature:Ambient
CAS Number:2369-89-3
Formula:C₈H₁₀N₄
MDL Number:MFCD00995337